DURAN® Funnels, Powder
Supplier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
213543804 213544603 213545102 213545505 213546107 213544106 213543307
221-1700EA 15.9 GBP
221-1700 221-1605 221-1702 221-1703 221-1704 221-1701 221-0281
DURAN® Funnels, Powder
Funnels Powder Funnels
DURAN®, borosilicate glass 3.3, clear.
  • Good chemical and physical properties
  • Use in applications that require good thermal resistance
  • Quick and easy to clean
With smooth internal walls. For transferring powdery materials. Can also be used for liquids. Funnel lip is ground and fire-polished.