Funnel for conical ground joints
Supplier: VITLAB
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
70594 70694 70494
221-1409EA 104 GBP
221-1409 221-1410 221-1418
Funnel for conical ground joints
Funnels Filtration Funnels Funnels for Conical Joints
PP, translucent.
  • Ideal for transferring liquid or powdered reagents to multiple-neck flasks
  • Very good chemical resistance
  • Autoclavable
The funnel body is flattened on the side and the end of the shaft is shaped to fit conical ground joints.