Funnel with ball and lid, electrostatic conductive, ARNOLD, V2.0
Supplier: S.C.A.T.
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
317621 317622 317623 317624 317625 317626 317627 317628 317629 317630 317635 317638 317634 317633 319002 317631 317650
221-0292EA 223 GBP
221-0292 SCTV317622 SCTV317623 221-0293 221-0294 SCTV317626 SCTV317627 SCTV317628 SCTV317629 221-0295 SCTV317635 221-0297 SCTV317634 SCTV317633 SCTV319002 SCTV317631 SCTV317650
Funnel with ball and lid, electrostatic conductive, ARNOLD, V2.0
Funnels Safety Funnels
The strong and electrically conductive safety funnel 'ARNOLD' V2.0 with lid and ball meets the highest requirements for safe disposal of hazardous liquid waste due to its combination of two independent safety mechanisms. The two safety features consist of a ball integrated in the funnel basin and a sealing ring incorporated in the hinged lid of the funnel, which prevent the escape of toxic gases. The funnel is especially designed for explosive or flammable substances, as the PE-HD material used is eclectically conductive and charges can be dissipated via the ground cable.
  • Electrically
More Product Information