Conical separating funnels
Supplier: JONES FR
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
WJ572 WJ573 WJ574 WJ575 WJ578 WJ579 WJ580 WJ581
221-0067EA 218 GBP
221-0067 221-0068 221-0069 221-0070 221-0071 221-0072 221-0073 221-0074
Conical separating funnels
Funnels Separating Funnels
Borosilicate glass, with NS-stopper.
With socket and plain stem. Available with interchangeable solid glass key or alternatively PTFE key for grease-free performance. Without graduation.