Separating funnels, pear shaped
Supplier: JONES FR
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
WJ559 WJ561 WJ562 WJ566 WJ567 WJ568 WJ569
221-0060EA 165 GBP
221-0060 221-0061 221-0062 221-0063 221-0064 221-0065 221-0066
Separating funnels, pear shaped
Funnels Separating Funnels
Borosilicate glass, with glass stopper.
With socket and plain stem. Available with interchangeable solid glass key or alternatively PTFE key for grease-free performance. Without graduation.