Dimethyl malonate 99+%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: 1,3-Propanedioic acid dimethyl ester, Malonic acid dimethyl ester

220700010 220702500 220700051
220700010.EA 63 GBP
220700010. 220702500. 220700051.
Dimethyl malonate 99+%
Dimethyl malonate
Formula: C₅H₈O₄
MW: 132.12 g/mol
Boiling Pt: 180…181 °C (760 mmHg)
Melting Pt: –62 °C
Density: 1.156 g/cm³
Flash Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD00008460
CAS Number: 108-59-8
EINECS: 203-597-8
Merck Index: 12,06169

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4120 to 1.4140 (20°C, 589 nm)
Water ≤0.1 % (K.F.)
GC ≥99.0 %
Free acid ≤0.1 % (as malonic acid)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR