Benzene ≥99.5% (by GC), AnalaR NORMAPUR® ACS analytical reagent

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Cyclohexatriene

21806.290EA 87.4 GBP
21806.290
Benzene ≥99.5% (by GC), AnalaR NORMAPUR® ACS analytical reagent
Benzene
Formula: C₆H₆
Boiling Pt: 80.1 °C (1013 hPa)
Melting Pt: 5.5 °C
Density: 0.878 g/cm³ (20 °C)
Flash Pt: –11 °C
Storage Temperature: Ambient
MDL Number: MFCD00003009
CAS Number: 71-43-2
EINECS: 200-753-7
UN: 1114
ADR: 3,II
REACH: 01-2119447106-44
Merck Index: 13,01066
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d2c0146712ab33142aeeb3843b02381e6bda4028&vid=cae270bfe08ae6ab4b14d9e1775b89a82cfeec1f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity (GC) min. 99.5%
Identity (IR) passes test
Appearance clear colorless liquid
Coloration (APHA) max. 10
Refractive index (20°C, 589 nm) 1.499 - 1.502
Alkalinity (meq/g) max. 0.0001
Acidity (meq/g) max. 0.0001
Residue on evaporation max. 0.001%
Water max. 0.05%
Substance darkened by H2SO4 passes test
Sulphur compounds (S) max. 0.005%
Thiophene (C4H4S) passes test
Copper (Cu) max. 0.00002%
Iron (Fe) max. 0.00005%
Lead (Pb) max. 0.00002%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR