Sodium iodide 99, reagent, Fluka™

Supplier: Honeywell Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Hydriodic acid sodium salt, Sodium monoiodide, Ioduril

217638-5G 217638-25KG
217638-5GEA 36.8 GBP
217638-5G HONC217638-25KG
Sodium iodide 99, reagent, Fluka™
Sodium iodide

  • ≥95% for general laboratory use.

Formula: NaI
MW: 149.89 g/mol
Boiling Pt: 1.304 °C (1.013 hPa)
Melting Pt: 662 °C
Density: 3.67 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003532
CAS Number: 7681-82-5
EINECS: 231-679-3
Merck Index: 13,08705

Order Now

Specification Test Results

Assay of I 83.8 - 85.4 %
Water (Karl Fischer) Max. 1.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR