Stoppers, rubber, Suba-Seal
Supplier: LEY RUBBER
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
TO/25/RED_U TO/29/RED_U TO/33/RED TO/45/RED_U TO/37/CREAM_U TO/57/CREAM TO/57/RED_U TO/41/RED_U TO/45/CREAM TO/17/RED TO/45/RED TO/25/CREAM TO/37/RED
217-0720EA 71.4 GBP
217-0720 217-0721 LEYRTO/33/RED 217-0723 217-0765 LEYRTO/57/CREAM LEYRTO/57/RED_U LEYRTO/41/RED_U LEYRTO/45/CREAM LEYRTO/17/RED LEYRTO/45/RED LEYRTO/25/CREAM LEYRTO/37/RED
Stoppers, rubber, Suba-Seal
Stoppers Rubber Stoppers
Turnover stoppers create a very effective double seal for narrow aperture containers.
  • No smell or taste
  • Will not yellow over time
  • Higher tear strength with less vulnerability to splitting or mechanical damage
External serrations create an excellent internal seal with the flexible turnover sleeve creating a secure seal on the container's outer neck. The head is easily pierced with a hypodermic needle.