Caps, BACTI-CAPS for reagent and culture tubes
Supplier: MHP Industries
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
BC/13/BK BC/13/W BC/13/R BC/13/Y BC/13/BU BC/13/G BC/16/BK BC/16/W BC/16/R BC/16/Y BC/16/BU BC/16/G BC/19/B BC/19/W BC/19/R BC/19/Y BC/19/BU BC/19/G BC/25/BK BC/25/W BC/25/R BC/25/B BC/25/G BC/38NATURAL BC/19/BK
217-0029EA 112 GBP
217-0029 217-0033 217-0032 217-0034 217-0030 217-0031 217-0035 217-0039 217-0038 217-0040 217-0036 217-0037 217-0041 217-0046 217-0045 217-0047 217-0043 217-0044 217-0049 217-0053 217-0052 217-0050 217-0051 217-0055 217-0042
Caps, BACTI-CAPS for reagent and culture tubes
Closures Test/Sample Tube Closures
PP, coloured.
  • Seal the tubes completely yet allow exchange of controlled gases
  • Excellent chemical resistance, autoclavable
  • Available colour-coded for tubes of different sizes
For smooth-edged glass or plastic reagent and culture tubes.
Ideal for cell culture. The alternative to conventional cotton and pulp stoppers.