Hydrogen peroxide 30% (w/w) in water contains inhibitor ACS

Supplier: Honeywell Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

216763-100ML 216763-500ML 216763-4L
216763-100MLEA 27.7 GBP
216763-100ML 216763-500ML 216763-4L
Hydrogen peroxide 30% (w/w) in water contains inhibitor ACS
Hydrogen peroxide
Formula: H₂O₂
Boiling Pt: Decomposes on heating
Melting Pt: -26 °C
Density: 1.11 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011333
CAS Number: 7722-84-1
UN: 2014
ADR: 5.1,II

Order Now

Specification Test Results

Assay 29.0 - 32.0 %
Non-volatile matter Max. 0.002 %
Titrable Acid Max. 0.0006 meq/g
Iron (Fe) Max. 0.5 ppm
Heavy metals (as Pb) Max. 1 ppm
Ammonium (NH₄) Max. 5 ppm
Chloride (Cl) Max. 2 ppm
Nitrate (NO₃) Max. 2 ppm
Phosphate (PO₄) Max. 2 ppm
Sulfate (SO₄) Max. 5 ppm
APHA Max. 10
Vapor Density 1 (vs air)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR