Carboys, narrow neck, PC, with screw cap, Nalgene®
Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
2251-0020 2251-0050
216-8781EA 308 GBP
216-8781 216-8782
Carboys, narrow neck, PC, with screw cap, Nalgene®
Bottles Carboys
PC, transparent with white PP screw cap.
  • Good chemical resistance
  • Temperature resistant from ‒100 to +135 °C, autoclavable
  • Light and robust, graduated
Can be used in any situation. Particularly suitable for the production of large quantities of nutrient and culture media, particularly for cultures that require light, e.g. algae. Ideal for storing and dispensing reagents and distilled water. Can be used in cold rooms and at temperatures below freezing.
Conduct large-volume media and culture preparation with these polycarbonate Nalgene Clearboys,More Product Information