Carboys, narrow neck, HDPE, with screw cap
Supplier: Kautex
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
350-84131 350-84132 350-84133 350-84135 350-84136 350-84137 350-84138 350-84139
216-5502EA 49.4 GBP
216-5502 216-5514 216-5504 216-5506 216-5508 216-5512 216-5516 216-5518
Carboys, narrow neck, HDPE, with screw cap
Bottles Carboys
HDPE. Natural colour. Blue litre scale.
  • Printed litre scale
  • Optional stopcock connection
  • Suitable for food
  • Halal certified
Screwed-on closure. Dust cap serves as a dust protection, but does not seal the carboy. With and without threaded stopcock connection.