Watch glasses
Supplier: SMITH SCIENTIFIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
170/150MM 170/170MM 170/190MM 170/1100MM 170/1150MM
216-4463EA 97.9 GBP
216-4463 216-4464 216-4466 216-4460 216-4461
Watch glasses
Watch Glasses
Soda-lime glass.
  • Good chemical and physical properties
  • Use in applications that do not involve high thermal stresses
With ground edges for covering beakers.