Dishes, flat bottom, with spout
Supplier: Agencinox
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
312A 316A 320A 325A 306 308 310 312 304
216-2715EA 62.5 GBP
216-2715 216-2716 216-2717 216-2718 216-2711 216-2712 216-2713 216-2714 AGEN304
Dishes, flat bottom, with spout
Trays/Dishes/Pots Laboratory Trays/Dishes
Stainless steel 18/10.
  • Without handles
  • Good chemical resistance
  • Excellent thermal resistance
Round multipurpose dishes for a wide range of uses.