Carboys, narrow neck, PC, square, Biotainer™, Nalgene®
Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
383405-16 383405-42 383410-08 383410-42 383423-42
216-2282EA 841 GBP
216-2282 216-2283 216-2284 216-2285 216-2286
Carboys, narrow neck, PC, square, Biotainer™, Nalgene®
Bottles Carboys
Specifically designed for pharmaceutical and biological manufacturing, Thermo Scientific Nalgene polycarbonate biotainer bottles and carboys reduce the risk of carry-over contamination.
  • Narrow mouth, square shape with handle
  • Sterile and ready to use
  • Translucent construction
  • Robust and versatile
  • Graduated
  • Not autoclavable
  • Closures are included
  • Liner material: Silicone
Durable, translucent PC construction ensures the integrity of your sample, even frozen to −100 °C. Sterile, ready-to-use products with a sterility assurance level of 10⁻⁶ inside theMore Product Information