Jerricans, wide neck, HDPE
Supplier: hünersdorff GmbH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
818200 818300
216-5220EA 113 GBP
216-5220 216-2117
Jerricans, wide neck, HDPE
Bottles Jerricans
HDPE, translucent.
  • Good chemical resistance, good temperature resistance
  • Sealable cap
  • All jerricans feature a screw thread and stoppers on the narrow side
The extra wide filling opening (88 mm I.D.) makes cleaning the inside much easier, which is particularly important when storing and transporting sensitive substances. The carrying handle and hinged screw cap ensure this product can be transported easily and securely. The design with a 10-litre capacity features an additional mini spout (18 mm I.D.). UV-stabilised. The 22-litre jerricans are stackable.