Laboratory trays
Supplier: KARTELL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
5701 5702 5703 5704 5705 5706 5707 5708 5709 5710 5712 5713 5714 5715
216-2101EA 34.4 GBP
216-2101 216-1684 216-2103 216-1683 216-2105 216-2106 216-4307 216-2108 216-2109 216-2110 216-2112 216-1681 216-2114 216-2115
Laboratory trays
Trays/Dishes/Pots Laboratory Trays/Dishes
PS, white.
  • Robust, impact resistant design
  • Heat resistant up to +70 °C
  • Smooth surfaces for easy cleaning
General purpose trays for a wide variety of laboratory uses. Also suitable for use in the food industry.