Infectious products shippers
Supplier: E3 Cortex
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
56CRYOPACK12L 56CRYOPACK4L 56CRYOPACK6L 371BIOT.25ST
216-2037EA 1080 GBP
216-2037 216-2039 216-2040 CORT371BIOT.25ST
Infectious products shippers
Mailers and Shipping Containers
Micro-fluted cardboard treated against moisture (Biotainer); outer double fluted carton (Cryopack®).
  • Cardboard wedge
  • Sheet of absorbent cloth
  • Watertight secondary PP container
These packages are designed for the transport of infected or contaminated consumables. They are suitable for applications in room temperature (Biotainer®). They can also be used in controlled temperature of 1 to 8 °C or in freezing temperatures with dry ice (Cryopack®).
Approved by the UN. Comply with international regulations in force IATA class 6.2 IMDG and ADR RIG.