VWR®, Carboys, Round, Sanitary Neck, PP
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
216-1897EA 216 GBP
216-1897 216-1910 216-1920
VWR®, Carboys, Round, Sanitary Neck, PP
Bottles Carboys
Ideal for bioprocessing applications. Features a sanitary flange (76 mm) that accepts standard TC fittings.
  • Convenient shoulder handles
  • Very good chemical resistance
  • Autoclavable
Certifications: Resin meets US FDA 21 CFR, USP Class VI.