Assortment Boxes, PP
Supplier: SEMADENI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1775 1771 1772 1774 1770 1773
215-0289EA 67.4 GBP
215-0289 216-1268 216-7331 216-7332 SEMA1770 SEMA1773
Assortment Boxes, PP
Boxes Storage Boxes
These assortment boxes, made of PP, have a transparent stable hinged lid with lock and tamper-evident seals.
  • Robust and strong design
  • All models can be stacked together
  • Lid is fixed on each tray so that the contents don't mix even if box is turned upside down