Laboratory trays, coloured
Supplier: MULTIROIR
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
39201PLBBLANC 39205PLBCBLEU 39204PLBCVERT 39206PLBROUGE 39101 39105PLACBLEU 39103 39104 39106
216-1125EA 27.1 GBP
216-1125 216-1127 216-1129 216-1130 216-1115 216-1117 216-1119 216-1121 216-1123
Laboratory trays, coloured
Trays/Dishes/Pots Laboratory Trays/Dishes
PS.
  • Five different colours
  • Two sizes
  • Good chemical and temperature resistance
Multipurpose trays for a wide range of applications.