VWR®, Crucibles, PTFE
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
216-0450EA 23.8 GBP
216-0450 216-0451 216-0452 216-0453
VWR®, Crucibles, PTFE
Crucibles
Isostatically moulded in pure PTFE, FDA approved.
  • Chemically resistant and inert
  • Crack-free
  • Withstand maximum temperature of 280 °C