Dishes, evaporating, silica vitreosil
Supplier: TECHNICAL QUARTZ SOLUTIONS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
73000 73001 73002 73004 73005 73009
TSLA73000EA 123 GBP
TSLA73000 216-0291 216-4468 216-4469 216-0292 TSLA73009
Dishes, evaporating, silica vitreosil
Trays/Dishes/Pots Evaporating Basins/Dishes
Basins with round bottom and spout made from quartz Vitreosil®.
  • Very good chemical resistance
  • Very good temperature resistance
  • Easy to clean