Stainless Steel Buckets
Supplier: BOCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
8315 8316 8317 8320 8322 8323 8324 8332 8333 8334 8321
216-1600EA 310 GBP
216-1600 216-1601 216-1602 216-0227 216-0228 216-0229 216-0230 216-0231 216-0232 216-0233 216-1603
Stainless Steel Buckets
Buckets
Buckets, stainless steel 18/10.
  • Very good chemical resistance, very good thermal resistance
  • High quality, very stable
  • Matching lids available optionally
For use in laboratories and industry. Graduated with handle. Seamless handle tabs and smooth surfaces for fast, easy cleaning.