VWR®, Dishes, Evaporating, Tall Form
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
216-0179EA 24.6 GBP
216-0179 216-0180 216-0181 216-0182 216-0183 216-0184 216-0185
VWR®, Dishes, Evaporating, Tall Form
Trays/Dishes/Pots Evaporating Basins/Dishes
PTFE, with spout.
  • Very good chemical resistance
  • Temperature resistant from –200 to +280 °C
  • Exceptionally smooth inner walls to prevent contamination
Thick walled, conical shape. Extremely robust.