Wash bottles, wide neck, Unitary™, Nalgene®
Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
2402-0125 2402-0250 2402-0500 2402-0750 2402-1000
215-8111EA 61.6 GBP
215-8111 215-8112 215-8113 215-8114 215-8115
Wash bottles, wide neck, Unitary™, Nalgene®
Bottles Wash Bottles
LDPE, translucent, with PP screw cap.
  • Wide neck for quick, safe filling, leakproof closure
  • Formed from a single piece, outlet tube can no longer become contaminated or leak
  • Jet tip can be cut to increase flow rate
  • Good chemical resistance, heat resistant up to 80 °C
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification IRECS; ISO 14001 (Certificate Registration No. EMS 752603)