Wash bottles, narrow neck, autoclavable, Nalgene®
Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
2405-0500 2405-1000
215-8108EA 148 GBP
215-8108 215-8107
Wash bottles, narrow neck, autoclavable, Nalgene®
Bottles Wash Bottles
PPCO, translucent, PP spray cap.
  • Very good chemical resistance, heat resistant up to +121 °C
  • Flexible, leakproof closure
  • With blue 'Autoclavable' label and printed size code
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification NAR Certificate of Retirement