Media bottles, square, sterile, Nalgene®
Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
2019-0030 2019-0060 2019-0125 2019-0250 2019-0500 2019-1000 2019-2000
215-6700EA 469 GBP
215-6700 391-7122 391-7123 391-7124 391-7125 391-7126 391-7127
Media bottles, square, sterile, Nalgene®
Bottles Cell Culture Bottles
PETG bottle with HDPE screw closure.
  • Leakproof closure, break-resistant and durable
  • Reduced permeability to carbon dioxide and oxygen
  • Bottles and closures are radiation sterilised and non-pyrogenic to eliminate costly washing, depyrogenation and autoclaving steps
  • Heat-shrink band around closure and neck provides tamper-evident seal
  • 2 L size has moulded in hand grips and a 53 mm white closure
An inexpensive alternative to glass bottles for storage and transport of media, these heavy walled, square, media bottles are transparent and graduated.