Bottles, clear or amber glass, DURAN® PURE
Supplier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
818062402 818063604 818061406 818011401 818012406 818013608 818014407 818015403 818016305 818017301 818018606 818019105 818064403 818065408 818066301 818067306 818068602 818069101
215-4765EA 344 GBP
215-4765 215-4766 215-4764 215-4755 215-4756 215-4757 215-4758 215-4759 215-4760 215-4761 215-4762 215-4763 215-4767 215-4768 215-4769 215-4770 215-4771 215-4772
Bottles, clear or amber glass, DURAN® PURE
Bottles Glass Bottles
DURAN® PURE bottles clear or amber, with GL thread and dust cover. DURAN® PURE is a range of primary packaging products dedicated to the specific requirements and needs of the pharmaceutical industry.
  • Made from DURAN® borosilicate glass 3.3 Type I
  • Protective dust cover is applied in-line at the end of the manufacturing process while the glass still has a very high level of cleanness
  • Lot specific certificate of conformance is available via 'Retrace Code' internet download (includes USP, Ph. Eur., JP analysis per lot)
  • 100% lot traceability of DURAN® PURE
More Product Information