VWR® Bottles, Wide Neck, Square, with Cap
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
215-4550EA 13.1 GBP
215-4550 215-4551 215-4552 215-4553 215-4554 215-4555 215-4556 215-4557 215-4558 215-4559 215-4560 215-4561 215-4562 215-4563 215-4564
VWR® Bottles, Wide Neck, Square, with Cap
Bottles Plastic Bottles
Wide neck square bottles available in HDPE or PP material.
  • Amber color HDPE bottle protects light sensitive materials
  • PP closure ensures leak-proof sealing
  • Square shape is ideal for storage of dry chemicals or powder
Resin compliance to US FDA and US and EU Pharmacopiea.
PP bottles are autoclavable.