Torque wrench sockets for screw caps GL 25, GL 32 and GL 45, DURAN®
Supplier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
292331301 292331326 292331342 292331901 292331926 292332803 292332828 292332844
215-4273EA 218 GBP
215-4273 215-4274 215-4275 215-4276 215-4277 215-4278 215-4279 215-4280
Torque wrench sockets for screw caps GL 25, GL 32 and GL 45, DURAN®
Wrenches
Applying the correct closure torque to DURAN® caps is critical to achieving an effective seal. Therefore, a range of eight specifically designed splined sockets that fit most of the DURAN® GL-threaded plastic closures can be used.
  • Compliance – enables accurate closure of DURAN® GL 25, 32 and 45 caps to the recommended torques
  • Ease of use – cap thread size and type is permanently engraved into the top surface
  • Hygienic design – suitable for cleanroom use
  • Full traceability – engraved lot-specific code for certification
  • Convenient – socket hole fits 'industry
More Product Information