WHEATON® Bottle, Wide Neck, Round, AC Type
Supplier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
W216997 W216998 W216999 W217000 W217001 W217002 W217004 W216995 W216996 W217003 W217007 W217009
215-4235EA 59.3 GBP
215-4235 215-4236 215-4237 215-4238 215-4239 215-4240 215-4242 215-4233 215-4234 215-4241 WHEAW217007 WHEAW217009
WHEATON® Bottle, Wide Neck, Round, AC Type
Bottles Glass Bottles
Bottles manufactured from clear soda lime glass.
  • Packaged in convenience packs with caps attached to maintain cleanliness
  • Hazardous communication label included with each case of bottles which allows for content ID and safety notification
  • Tamper evident seal included with each case of bottles keeps bottle sealed which protecting contents from accidental misuse
Wide mouth bottles are ideal for dry and viscous samples.