Dropping bottles, LDPE, Wheaton®
Supplier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
W242831 W242825 W242822 W242822-A W242823 W242823-A W242824 W242825-A W242826 W242826-A W242827 W242824-A W242821-A W242821 W242834-A W242831-A W242832 W242832-A W242833 W242833-A W242834 W242835 W242835-A W242836 W242836-A W242837
215-4136EA 130 GBP
215-4136 215-4145 215-4139 WHEAW242822-A 215-4141 WHEAW242823-A 215-4143 WHEAW242825-A 215-4147 WHEAW242826-A 215-4149 WHEAW242824-A WHEAW242821-A 215-4137 WHEAW242834-A WHEAW242831-A 215-4138 WHEAW242832-A 215-4140 WHEAW242833-A 215-4142 215-4144 WHEAW242835-A 215-4146 WHEAW242836-A 215-4148
Dropping bottles, LDPE, Wheaton®
Bottles Dropping Bottles
Dropping bottles made of natural or white LDPE. The white LDPE bottles tends to protect the sensitive samples from UV light.
  • Good chemical resistance
  • Good temperature resistance