Dropper tip caps, WHEATON®
Supplier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
242502 242504 242506 242508 242510 242512 242514 242516 242518 242520 242522 242524 242526 242528 242530 242501 242517 242503 242507 242513 242532 242533 242534 242535 242536 242537 242538 242540 242505 242511 242515 242521 242531 242523 242525 242527 W242502-A W242503-A W242504-A W242508-A W242510-A W242522-A W242523-A W242525-A W242512-A W242514-A W242515-A W242520-A W242521-01-A
215-4005EA 33.9 GBP
215-4005 215-4007 215-4009 215-2763 215-4011 215-2764 215-4014 215-4016 215-4018 215-4019 215-4021 215-2765 215-4024 215-4026 215-4027 215-4004 215-4017 215-4006 215-4010 215-4013 215-4029 215-4030 215-4031 215-4032 215-4033 215-4034 215-4035 215-4036 215-4008 215-4012 215-4015 215-4020 215-4028 215-4022 215-4023 215-4025 215-4037 215-4038 215-4039 215-4040 215-4041 WHEAW242522-A 215-4045 215-4046 WHEAW242512-A WHEAW242514-A 215-4042 215-4043 215-4044
Dropper tip caps, WHEATON®
Closures Dropper Caps
Screw cap made of radiation grade polypropylene.
  • Very good chemical resistance
  • Very good temperature resistance
  • Variety of colors