VWR®, Dropping Bottles
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
215-3816EA 39.8 GBP
215-3816 215-3817 215-3818 215-3819 215-3820 215-3821 215-3822 215-3823 215-3824 215-3825 215-3826 215-3827 215-3828
VWR®, Dropping Bottles
Bottles Dropping Bottles
Soda-lime glass, amber.
  • Good chemical and physical properties
  • Pipette kit not included
Amber to protect light-sensitive substances, with DIN 18 thread.