Screw caps, DURAN® PURE Premium
Supplier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
891088679 891081306 898862802 891088678
215-3740EA 16.2 GBP
215-3740 215-3739 215-4773 SCOT891088678
Screw caps, DURAN® PURE Premium
Closures Bottle Closures
Premium caps are produced and assembled with previously specially cleaned PTFE coated silicone seals. Made from Fluoropolymer (PFA - TpCh260TZ).
  • Outstanding temperature resistance from –196 to + 200 °C
  • Very high chemical resistance
  • High leak-tightness through use of PTFE coated silicone seal
  • No dye additives (leaching of dyes is avoided)
Order Now
SPECIFICATIONS
Environmentally Preferable