Bottles, glass
Supplier: Azpack
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
215-2927 215-2926 215-2928 215-2929 215-2930 215-2931 215-2932 215-2933 215-2934 215-2935 215-2936 215-2937 215-2938
215-2927EA 94.2 GBP
215-2927 215-2926 215-2928 215-2929 215-2930 215-2931 215-2932 215-2933 215-2934 215-2935 215-2936 215-2937 215-2938
Bottles, glass
Bottles Glass Bottles
Lightweight glass bottles made from type III soda lime glass in either white flint or amber. Fitted with caps as indicated. These bottles have a narrow neck and are ideal for storing liquids.