Bottles, wide neck, PP, round, with screw cap, AZLON®
Supplier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
BLP0150P BWP0250P BWP0500P BWP1000P BWP2000P BLP0100P BLP0125P BWP0030PN BWP0060PN
215-0390EA 0 GBP
215-0390 215-0392 215-0393 215-0394 215-0560 215-2869 215-2870 215-2871 215-0552
Bottles, wide neck, PP, round, with screw cap, AZLON®
Bottles Plastic Bottles
PP, high clarity, rigid with PP screw caps.
  • Leakproof, liner-less design
  • Smooth inner contours for efficient ease of pouring
  • Heavy-duty and durable
  • Good chemical resistance, autoclavable at +121 °C
Ideal for a wide range of liquid storage, media preparation and sampling applications.
Caution: When autoclaving bottles always ensure the caps are completely loosened or removed to prevent accidental implosion.