DURAN® Reactor, Stirred, GLS 80 Thread
Supplier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1200379 1200380 1200381 1200382 1200385 1200386 1200383 212639107
215-2468EA 348 GBP
215-2468 441-0550 441-0551 215-2473 215-2469 215-2470 215-2474 215-4158
DURAN® Reactor, Stirred, GLS 80 Thread
Flasks Spinner Flasks
DURAN®, borosilicate glass 3.3, clear, PP, PTFE, PEEK, stainless steel.
  • Parts in contact with media conform fully to FDA requirements
  • The agitating element is exchangeable
  • With the aid of components from the DURAN® connection system extra media bottles can be connected or a sterile pressure release is possible
The GLS 80 stirred reactor is suitable for a wide range of laboratory mixing processes. The connections provided permit addition or removal of media from the bottle during the mixing process. The whole assembly can be autoclaved and is thereforeMore Product Information