VWR®, Bottles, Narrow Neck, with Screw Cap
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
215-2327EA 62.1 GBP
215-2327 215-2329 215-2328 215-2330 215-2337 215-2338 215-2325 215-2326 215-3275
VWR®, Bottles, Narrow Neck, with Screw Cap
Bottles Glass Bottles
Amber borosilicate 3.3 glass with PP screw cap and pouring ring.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
The economical alternative for a wide range of laboratory applications. Round, with graduations, DIN GL 45 thread, pouring ring, and blue PP screw cap. Amber coloured to protect light-sensitive materials.
Purely external application of the colour ensures unchanged glass properties on the inner surface of the bottles.
Certifications: ISO 4796