Bottles, narrow neck, alpha, lightweight with screw caps
Supplier: Azpack
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
215-2100 215-2101 215-2102 215-2103 215-2104 215-2105 215-2106 215-2107 215-2108 215-2109 215-2110 215-2111 215-2112 215-2113 215-2114 215-2115 215-2117 215-2118 215-2119 215-2120 215-2121 215-2122 215-2123 215-2124 215-2125 215-2126 215-2127 215-2128 215-2129 215-2130
215-2100EA 113 GBP
215-2100 215-2101 215-2102 215-2103 215-2104 215-2105 215-2106 215-2107 215-2108 215-2109 215-2110 215-2111 215-2112 215-2113 215-2114 215-2115 215-2117 215-2118 215-2119 215-2120 215-2121 215-2122 215-2123 215-2124 215-2125 215-2126 215-2127 215-2128 215-2129 215-2130
Bottles, narrow neck, alpha, lightweight with screw caps
Bottles Glass Bottles
Clear or amber lightweight glass.
  • For use in applications without high thermal loads
  • Amber coloured to protect light-sensitive material
  • A lighter, stronger and more uniform range of bottles
Large range of standard round glass bottles. All sizes of the lightweight bottles have exactly the same neck which means that only one cap size for the entire range has to be stocked.