Bottles, narrow neck, with standard ground stoppers
Supplier: Kavalier
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
N632414126050 N632414126500 N632414226250 N632414226950
215-1750EA 21.2 GBP
215-1750 215-1753 215-1760 215-1763
Bottles, narrow neck, with standard ground stoppers
Bottles Glass Bottles
Soda-lime glass, clear or amber, with standard ground joint and glass stopper.
  • Good chemical and physical properties
  • For use in applications without high thermal loads
  • Solid, replaceable glass stoppers
Amber to protect light-sensitive materials.
DIN 12036, ISO 4796
Certifications: DIN 12036, ISO 4796