Bottles, wide neck, with standard ground stoppers
Supplier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
231852404 231853606 231854405 231855401 231882407 231883609 231884408 231885404
215-1019EA 15.4 GBP
215-1019 215-1023 215-1737 215-1478 215-1479 215-1480 215-1481 215-1015
Bottles, wide neck, with standard ground stoppers
Bottles Glass Bottles
Soda-lime glass, clear or brown, with standard ground joint and glass stopper.
  • Good chemical and physical properties
  • For use in applications that are not associated with high thermal loads
  • Stable and tough
Available in crystal clear glass or brown tinted to protect light-sensitive substances.
Certifications: DIN 12039