Bottles, wide neck, with screw cap
Supplier: LAMAPLAST
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
413 412 411 410 409 408
215-1364EA 8.8 GBP
215-1364 215-0914 215-0913 215-0912 215-0911 215-0910
Bottles, wide neck, with screw cap
Bottles Plastic Bottles
LDPE, translucent.
  • Good chemical resistance, heat resistant up to 70 °C
  • Leakproof
  • Graduated
  • Suitable for food contact
Double closure cap, shoulder and perforated cap to allow the installation of seals and labels to ensure the integrity of the contents. Can be used anywhere in the laboratory and for sampling. Wide mouth for rapid filling and dispensing of powders, pastes and liquids.