Bottles, wide neck, with screw cap, AZLON®
Supplier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
BWP0060AP BWP0150AP BWP0250AP BWP0500AP BWP1000AP BWP2000AP
215-0551EA 0 GBP
215-0551 215-0553 215-0554 215-0556 215-0558 215-0561
Bottles, wide neck, with screw cap, AZLON®
Bottles Plastic Bottles
PP, amber, opaque, rigid bottles with black PP screw caps.
  • Leakproof, liner-less design
  • Smooth inner contours for efficient pouring and ease of cleaning
  • Heavy-duty and durable
  • Good general chemical resistance, autoclavable at +121 °C
Ideal for sample collection and storage of light-sensitive materials. Wide opening for rapid filling and dispensing of powders, pastes and liquids.
Caution: When autoclaving bottles always ensure the caps are completely loosened or removed to prevent accidental implosion.