Bottles, aluminium, with screw cap
Supplier: BURKLE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
0327-0120 0327-1200 0327-0060 0327-0600 0327-0300 0327-0038 0327-3000 0327-5600
215-0260EA 4.2 GBP
215-0260 215-0262 215-4285 215-4286 215-0261 215-4284 215-4287 215-4288
Bottles, aluminium, with screw cap
Bottles Metal Bottles
Pure aluminium (material AL 99,5). This aluminium bottle is particularly suitable for storing, transport, or sending substances and reference samples which have to be stored completely unaltered.
  • With tamper-evident screw cap made of PP with aluminium inner seal
  • Suitable for food (exception: highly acidic and salt-containing foods)
  • With UN approval (for 60 to 5600 ml)
  • Can be completely emptied without residues
  • Odour-neutral
  • Impervious to light, rustproof