Bottles, narrow neck, round, LDPE, with screw cap
Supplier: LAMAPLAST
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
326G 326 324G 325 324 323G 323 319 317 325G
LAMA326GEA 8.9 GBP
LAMA326G 215-0166 LAMA324G 215-0149 215-0148 LAMA323G 215-0147 215-0146 215-0145 LAMA325G
Bottles, narrow neck, round, LDPE, with screw cap
Bottles Plastic Bottles
LDPE, translucent or grey.
  • Leakproof, suitable for foodstuffs
  • Tagging points on shoulder and cap enables use of tamper evident tags and seals
  • Easy grip cap
  • Grey coloured for photosensitive products
Insert in cap ensures a good seal. Sloping shoulders reduce tendency for any residual liquids when pouring and emptying.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904