Diphosphorus pentoxide ≥98.0%, GPR RECTAPUR®

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

21410.293EA 95.9 GBP
21410.293
Diphosphorus pentoxide ≥98.0%, GPR RECTAPUR®
di-Phosphorus pentoxide
Formula: P₂O₅
Melting Pt: 562 °C
Density: 2.253 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011440
CAS Number: 1314-56-3
EINECS: 215-236-1
UN: 1807
ADR: 8,II
Merck Index: 13,07441

Order Now

Specification Test Results

Assay Min. 98.0%
Heavy metals (as Pb) Max. 100 ppm
Reducing substances (as P₂O₃) Max. 0.02 %
Cl (Chloride) Max. 50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR