Flasks acc. to Kjeldahl, narrow neck, with flared rim, DURAN®
Supplier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
212311707 212312403 212313605 212314404 212315109 212315409
214-4105EA 27.4 GBP
214-4105 214-4110 214-4115 214-4120 214-4122 214-4125
Flasks acc. to Kjeldahl, narrow neck, with flared rim, DURAN®
Flasks Kjeldahl Flasks
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
With printed indication of contents and white labelling area.
Certifications: DIN 12360